Learn More
Sciteck™ SVT™ Adulteration SAM-Set™ Calibrator Series
For use in validity tests for pH, creatinine, nitrite, and refractometer (specific gravity)
Manufacturer:  SCITECK DIAGNOSTICS INC A41040
 View more versions of this product
Catalog No. 23-424-4575
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
SAM-Set™ calibrator series refractometer 1.0000
600nm
Refractometer (specific gravity), pH meter (pH), automated clinical chemistry analyzer
2° to 10°C
Human Urine
Forensic/toxicology use only
CAP FDT the Gold Standard, SAMHSA, HHS CLIA
60mL