Sciteck™ SVT™ Adulteration Specific Gravity Reagent

For in vitro diagnostic use to determine specific gravity of solution

Manufacturer: SCITECK DIAGNOSTICS INC 1185

 View more versions of this product

Catalog No. 23-424-4528

  Check Availability
Add to Cart

Disclaimers:

U.S. and Foreign Patents Pending: 5,753,451.


    Specifications

    Product Type SVT™ specific gravity calibrator 1.005
    Wavelength 600nm
    For Use With (Application) In-vitro diagnostic use to determine the specific gravity of a solution, forensic/toxicology use
    Storage Requirements 2° to 10°C
    Quantity 60mL,60mL
    Source Human Urine

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit