Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Sciteck™ UR-N-Control™ Tri-Level Adulteration QC Set
For use with automated analyzer systems performing adulteration detection
Manufacturer:  SCITECK DIAGNOSTICS INC 1191
 View more versions of this product
Catalog No. 23-424-4560
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Tri-level adulteration QC set
Synthetic Urine matrix containing Urea, Creatinine, Phosphates, Sodium, Chloride, Potassium, and other normal Urinary constituents
9 to 11
Automated analyzer system
125mL
Human Urine
500 to 1000mg/L
Performing adulteration testing and Urinalysis
2° to 10°C