Learn More
Sciteck™ UR-N-Control™ Tri-Level Adulteration QC Set
For use with automated analyzer systems performing adulteration detection
Manufacturer:  SCITECK DIAGNOSTICS INC 1192
 View more versions of this product
Catalog No. 23-424-4561
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Tri-level adulteration QC set
Synthetic Urine matrix containing Urea, Creatinine, Phosphates, Sodium, Chloride, Potassium, and other normal Urinary constituents
5.5 to 7.5
Automated analyzer system
125mL
Human Urine
250 to 750mg/L
Performing adulteration testing and Urinalysis
2° to 10°C