Sciteck™ UR-N-Control™ Tri-Level Adulteration QC Set

For use with automated analyzer systems performing adulteration detection

Manufacturer: SCITECK DIAGNOSTICS INC 1192NP

 View more versions of this product

Catalog No. 23-424-4562

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type Tri-level adulteration QC set
    Source Human Urine
    Includes Synthetic Urine matrix containing Urea, Creatinine, Phosphates, Sodium, Chloride, Potassium, and other normal Urinary constituents
    Concentration 0 to 0mg/mL
    pH 5.5 to 7.5
    For Use With (Application) Performing adulteration testing and Urinalysis
    For Use With (Equipment) Automated analyzer system
    Storage Requirements 2° to 10°C
    Quantity 125mL

    • In conjunction with drugs-of-abuse analysis on urine specimens
    • Made of synthetic urine matrix containing urea, creatinine, phosphates, sodium, chloride, potassium, and other normal urinary constituents
    • Free of potential contamination by hepatitis and HIV viruses
    • Yields a range of values for creatinine, pH, specific gravity and nitrite (oxidant)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit