Learn More
Sciteck™ UR-N-Control™ Tri-Level Adulteration QC Set
For use with automated analyzer systems performing adulteration detection
Manufacturer:  SCITECK DIAGNOSTICS INC 1193
 View more versions of this product
Catalog No. 23-424-4563
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Tri-level adulteration QC set
Synthetic Urine matrix containing Urea, Creatinine, Phosphates, Sodium, Chloride, Potassium, and other normal Urinary constituents
2.4 to 4.4
Automated analyzer system
125mL
Human Urine
0 to 250mg/L
Performing adulteration testing and Urinalysis
2° to 10°C