Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCL/Tal1 Mouse anti-Human, Clone: BTL73, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30762510UG
Description
SCL/Tal1 Monoclonal specifically detects SCL/Tal1 in Human samples. It is validated for Western Blot, ELISA.Specifications
SCL/Tal1 | |
Monoclonal | |
0.2 mg/ml | |
Western Blot 1-2 ug/ml, ELISA | |
BHLHA17, bHLHa17tal-1, Class A basic helix-loop-helix protein 17, SCLT-cell acute lymphocytic leukemia protein 1, Stem cell protein, TAL-1, T-cell acute lymphocytic leukemia 1, T-cell leukemia/lymphoma protein 5, TCL5tal-1 product | |
Recombinant human full-length SCL/Tal1 protein | |
100 μg | |
Phospho Specific | |
6886 | |
Store at 4°C. | |
IgG1 κ |
Western Blot, ELISA | |
BTL73 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title