Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCL/Tal1 Mouse anti-Human, Clone: BTL73, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen SCL/Tal1
Clone BTL73
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, ELISA
Applications Western Blot, ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122724
View Documents
Novus Biologicals
NBP30762520UG
20 μg
Each for $138.37
Add to cart
 
NB122723
View Documents
Novus Biologicals
NBP30762510UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

SCL/Tal1 Monoclonal specifically detects SCL/Tal1 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

SCL/Tal1
0.2 mg/ml
Western Blot, ELISA
Purified
RUO
Human
BHLHA17, bHLHa17tal-1, Class A basic helix-loop-helix protein 17, SCLT-cell acute lymphocytic leukemia protein 1, Stem cell protein, TAL-1, T-cell acute lymphocytic leukemia 1, T-cell leukemia/lymphoma protein 5, TCL5tal-1 product
Recombinant human full-length SCL/Tal1 protein
Monoclonal
Protein A or G purified
BTL73
Western Blot 1-2 ug/ml, ELISA
Unconjugated
Mouse
Phospho Specific
10 mM PBS with 0.05% BSA
6886
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit