Learn More
SCL/Tal1 Mouse anti-Human, Clone: C.15200012, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259207
Catalog No. NBP259207
Add to cart
Description
Description
SCL/Tal1 Monoclonal antibody specifically detects SCL/Tal1 in Human samples. It is validated for Chromatin Immunoprecipitation.
Specifications
Specifications
SCL/Tal1
C.15200012
Chromatin Immunoprecipitation 2 - 5 ug, Immunofluorescence 1:1000
Affinity Purified
TAL1
The exact immunogen is propietary information.
Affinity purified
RUO
Primary
6886
ChIP assay, Immunofluorescence
Unconjugated
PBS with 0.05% Sodium Azide
BHLHA17, bHLHa17tal-1, Class A basic helix-loop-helix protein 17, SCLT-cell acute lymphocytic leukemia protein 1, Stem cell protein, TAL-1, T-cell acute lymphocytic leukemia 1, T-cell leukemia/lymphoma protein 5, TCL5tal-1 product
Mouse
IgG1
50 ug
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Monoclonal
Human