Learn More
Crystal Chem IncSupplier Diversity Partner Sclerostin ELISA Kit, 96-wells DFS Item
Manufacturer:  Crystal Chem IncSupplier Diversity Partner 80735
The Sclerostin ELISA is a highly sensitive assay that is used to quantify levels of human sclerostin in serum, plasma, and urine. Highlights: Kit uses sample size of only 125 µL. High sensitivity of 54 pg/mL. Dynamic range of 125- 4000 ng/mL. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7853