Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCLY Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297263PCP
Description
SCLY Polyclonal antibody specifically detects SCLY in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
SCLY | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
EC 4.4.1.16, SCLhSCL, selenocysteine lyase | |
Produced in rabbits immunized with E. coli-derived Human SCLY fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
51540 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction