Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SCMH1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521804
Description
Recommended positive controls: 293T, H1299, HepG2, Raji. Predicted reactivity: Mouse (96%), Rat (97%), Xenopus laevis (87%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Associates with Polycomb group (PcG) multiprotein complexes; the complex class is required to maintain the transcriptionally repressive state of some genes. [UniProt].
Specifications
SCMH1 | |
Polyclonal | |
Unconjugated | |
SCMH1 | |
AI315320; AI851618; Polycomb protein SCMH1; scmh1; Scml1; Scml3; sex comb on midleg 1; sex comb on midleg homolog 1; sex comb on midleg homolog 1 (Drosophila); sex comb on midleg-like 1; sex comb on midleg-like 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22955 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q96GD3 | |
SCMH1 | |
Recombinant fragment corresponding to a region within amino acids 29 and 285 of SCMH1 (Uniprot ID#Q96GD3). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction