Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scn2b Mouse anti-Human, Mouse, Rat, Clone: S395-68, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259317

Catalog No. NBP259317


Add to cart

Description

Description

Scn2b Monoclonal antibody specifically detects Scn2b in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Scn2b
S395-68
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Purified
SCN2B
Fusion protein amino acids 1-215 (full-length) of mouse Nav beta2
Protein G purified
RUO
Primary
6327
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Unconjugated
PBS pH7.4, 50% glycerol with 0.1% Sodium Azide
neuronal voltage-gated sodium channel beta 2 subunit, sodium channel beta 2 subunit, sodium channel subunit beta-2, sodium channel, voltage-gated, type II, beta, sodium channel, voltage-gated, type II, beta polypeptide
Mouse
IgG2b
100 ug
Store at -20C.
Monoclonal
Detects 40kDa. Does not cross-reat with NavBeta1, Navbeta3, or Navbeta4.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only