Learn More
Scn2b Mouse anti-Human, Mouse, Rat, DyLight 488, Clone: S395-68, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259317G
Catalog No. NBP259317G
Add to cart
Description
Description
Scn2b Monoclonal antibody specifically detects Scn2b in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Scn2b
S395-68
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
SCN2B
Fusion protein amino acids 1-215 (full-length) of mouse Nav beta2
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 40kDa. Does not cross-reat with NavBeta1, Navbeta3, or Navbeta4.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
neuronal voltage-gated sodium channel beta 2 subunit, sodium channel beta 2 subunit, sodium channel subunit beta-2, sodium channel, voltage-gated, type II, beta, sodium channel, voltage-gated, type II, beta polypeptide
Mouse
IgG2b
100 ul
Primary
6327
Human, Mouse, Rat