Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCN3B Monoclonal Antibody (S396-29), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA527618
Description
SCN3B Monoclonal antibody specifically detects SCN3B in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
SCN3B | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4 | |
Q8BHK2, Q9JK00, Q9NY72 | |
Scn3b | |
Fusion protein amino acids 1-215 (full-length) of rat NavBeta3 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |
Immunocytochemistry, Western Blot | |
S396-29 | |
Unconjugated | |
Scn3b | |
1110001K16Rik; 4833414B02Rik; ATFB16; brain and heart sodium channel beta 3 subunit; BRGDA7; HSA243396; KIAA1158; LOC540925 protein; Scn3b; Scnb3; sodium channel beta 3 subunit; sodium channel subunit beta-3; sodium channel, voltage gated, type III beta subunit; sodium channel, voltage-gated, type III, beta; sodium channel, voltage-gated, type III, beta subunit; sodium voltage-gated channel beta subunit 3; voltage-gated sodium channel beta-3 subunit | |
Mouse | |
Protein G | |
RUO | |
235281, 245956, 55800 | |
-20°C | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction