Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCN3B Mouse anti-Human, Mouse, Rat, DyLight 550, Clone: S396-29, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259318R
Description
SCN3B Monoclonal specifically detects SCN3B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
SCN3B | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SCN3B | |
Fusion protein amino acids 1-215 (full-length) of rat NavBeta3 | |
100 μL | |
55800 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
S396-29 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
HSA243396, KIAA1158, SCNB3, sodium channel subunit beta-3, sodium channel, voltage-gated, type III, beta, voltage-gated sodium channel beta-3 subunit | |
Mouse | |
Protein G purified | |
Primary | |
Detects 40kDa. Does not cross-reat with NavBeta1, Navbeta2, or Navbeta4. | |
Store at 4C in the dark. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title