Learn More
SCN3B Mouse anti-Human, Mouse, Rat, DyLight 650, Clone: S396-29, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259318C
Catalog No. NBP259318C
Add to cart
Description
Description
SCN3B Monoclonal antibody specifically detects SCN3B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
SCN3B
S396-29
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
SCN3B
Fusion protein amino acids 1-215 (full-length) of rat NavBeta3
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 40kDa. Does not cross-reat with NavBeta1, Navbeta2, or Navbeta4.
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
HSA243396, KIAA1158, SCNB3, sodium channel subunit beta-3, sodium channel, voltage-gated, type III, beta, voltage-gated sodium channel beta-3 subunit
Mouse
IgG2b
100 ul
Primary
55800
Human, Mouse, Rat