Learn More
Aviva Systems Biology Corp SCO1 Antibody - middle region (ARP56592_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56592_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCO1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SCO1. Alternate names: SCOD1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-61917