Learn More
3M™ Scotch™ Box Sealing Tape
Scotch™ Box Sealing Tape is a high performance packing tape that securely closes a wide variety of heavy weight box materials, including recycled fiberboard.
Manufacturer:  3M™ 00021200730801
 View more versions of this product
Catalog No. 19-072-029
Add to cart
Description
Description
  • Polypropylene film backing is easy to unwind and resists abrasion, moisture and scuffing.
  • Backing easily conforms around edges and on rough surfaces for a tight seal.
  • High performance packaging tape for heavy duty applications on a wide range of materials
  • High edge tear and split resistance ideal for ecommerce and single parcel shipments
  • Polypropylene backing with synthetic rubber adhesive delivers conformability under stress
  • Easy unwind and smooth dispensing works well on high-speed automated packaging lines
  • Resists abrasion, moisture, chemicals and scuffing for long-lasting performance
  • Excellent adhesion to recycled liner paper and fiberboard
Specifications
Specifications
Clear
72 mm x 100 m
Hot Melt Synthetic Rubber Resin
Hot Melt Synthetic Rubber Resin
Box Sealing Tape
Automated Carton Sealing, Box Closure, Branding
24/case