Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Scotch™ Box Sealing Tape

Scotch™ Box Sealing Tape is a high performance packing tape that securely closes a wide variety of heavy weight box materials, including recycled fiberboard.

$424.80

Specifications

Adhesive Type Hot Melt Synthetic Rubber Resin
Color Clear
For Use With (Application) Automated Carton Sealing, Box Closure, Branding
Material Hot Melt Synthetic Rubber Resin
Type Box Sealing Tape
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Format Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Format Price Quantity & Availability  
19-072-063
View Documents
3M™
7010312313
48/case 36 mm x 50 m
Case of 48 for $424.80
Add to cart
 
19-072-028
View Documents
3M™
00021200730795
36/case 48 mm x 100 m This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-072-030
View Documents
3M™
00021200696053
24/case 72 mm x 50 m This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-072-029
View Documents
3M™
00021200730801
24/case 72 mm x 100 m This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Polypropylene film backing is easy to unwind and resists abrasion, moisture and scuffing.
  • Backing easily conforms around edges and on rough surfaces for a tight seal.
  • High performance packaging tape for heavy duty applications on a wide range of materials
  • High edge tear and split resistance ideal for ecommerce and single parcel shipments
  • Polypropylene backing with synthetic rubber adhesive delivers conformability under stress
  • Easy unwind and smooth dispensing works well on high-speed automated packaging lines
  • Resists abrasion, moisture, chemicals and scuffing for long-lasting performance
  • Excellent adhesion to recycled liner paper and fiberboard
Specifications

Specifications

Hot Melt Synthetic Rubber Resin
Automated Carton Sealing, Box Closure, Branding
Box Sealing Tape
Clear
Hot Melt Synthetic Rubber Resin
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit