Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Scotch™ Glass Cloth Electrical Tape
Scotch™ Glass Cloth Electrical Tape is premium grade and edge tear resistant.
Supplier: 3M™ 7000145501
Description
- Printable
- Non corrosive adhesive
- Offers solvent resistant protection
- Flame retardant and suitable for 600V rated, dry location application motors and transformers.
- Thermosetting, silicon adhesive provides an increased bond at high temperatures
- Glass cloth backing optimally resists resist shrinking and burning
- Used as a coil cover, anchor, banding, core, layer and crossover insulation
- Printable and provides high mechanical strength and excellent performance by withstanding high temperatures
- Withstands a temperature range of 32 to 392°F (0 to 200°C)
- UL Listed, Mil-I-19166C, CSA Certified and RoHS 2011/65/EU Compliant
Specifications
White | |
Glass Cloth Tape | |
69-1/2″X66′ |
Silicone | |
Automotive, Commericial Solutions, Communications, Consumer, Design and Construction, Electronics, Energy, Health Care, Manufacturing, Mining, Oil and Gas | |
66 ft. x 1/2 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction