Learn More
3M™ Scotch™ Industrial Grade Packing Tape
Scotch™ Industrial Grade Packing Tape securely closes a wide variety of medium weight box materials, including recycled fiberboard
Manufacturer:  3M™ 00021200192807
 View more versions of this product
Catalog No. 19-072-020
Add to cart
Description
Description
  • Industrial packaging tape for medium-duty applications on a wide range of materials
  • High edge tear and split resistance ideal for general industrial applications
  • Polypropylene backing with synthetic rubber adhesive delivers conformability under stress
  • Easy unwind and smooth dispensing works well on high-speed automated packaging lines
  • Resists abrasion, moisture, chemicals and scuffing for long-lasting performance
  • Excellent adhesion to recycled liner paper and fiberboard
  • Polypropylene film backing is easy to unwind and resists abrasion, moisture and scuffing.
  • Backing easily conforms around edges and on rough surfaces for a tight seal.
Specifications
Specifications
Industrial grade
Box Sealing Tape
Automated Carton Sealing, Box Closure, Branding
24 per case
Clear
72 mm x 100 m
Hot Melt Synthetic Rubber Resin
Hot Melt Synthetic Rubber Resin