Learn More
3M Company Scotch-Weld™ Epoxy Adhesive
3M Company Scotch-Weld™ Epoxy Adhesive is a flexible, fast-setting, two-part epoxy adhesive that cures clear for invisible bond lines
Manufacturer:  3M Company 62328714357
Catalog No. 19-070-917
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Maintains high shear and peel strength
  • Flexible when cured, making it a good choice for bonding dissimilar surfaces
  • 5 minute work life with handling strength in approximately 20 minutes at room temperature
  • Versatile bonder for metals, plastics, glass, ceramics and composites
  • Flexible adhesive formula provides strong, permanent bond even under vibration and impact
Specifications
Specifications
Safety
Structural Adhesives
Hard disk drive component assembly; Assembly
Clear
50 ml
12