Learn More
3M™ ScotchBlue™ Painter's Tape Display
3M™ ScotchBlue™ Painter's Tape Display is a medium adhesion tape that is ideal for painted walls and trim, woodwork, glass and metal.
Supplier: 3M™ 7010295728
Description
- Ideal for use on smooth or lightly textured walls, trim, glass and metal
- Sharp paint lines on a variety of surfaces
- Removes cleanly up to 14 days with no sticky residue, even in direct sunlight
- Medium adhesion level removes easily, while protecting your surfaces
- Indoor or outdoor use
- For ideal paint results make sure the surface is clean, dry and dust-free so that the tape sticks properly. Next apply tape onto the surface while pressing down as you go, then secure the tape by pressing down on it firmly with a smooth edge. And lastly let the tape set for about 30 to 60 minutes before painting. Wait until the paint is dry to the touch before removing the tape. Lift it up by slowly pulling it back on itself, then removing at a 45 degree angle.
Specifications
Blue | |
Decorating, Organizing and Crafts, DIY, Home Improvement, Home, Office, School Supplies | |
108 Rolls/Display |
Painter's Tape | |
2090-24B-N | |
60 yd. x 94 in. (54.8 x m x 24 mm) |