Scott™ Air-Pak™ 4.5 SCBA
Manufacturer: Scott Health & Safety 805609-0102
 View more versions of this product
Catalog No. 19-120-1408
  Check Availability
Add to Cart
    Specifications
    Pressure 4500psi
    Product Type Carbon Cylinder
    Duration 30 min.
    Includes Av-3000™ facepiece, e-z flo™ pressure demand regulator, vibralert™ low-air alarm; lightweight, lumbar supported backpack, heads up display (hud) and a rapid intervention crewith universal air connectio
    Material Aluminum Alloy Inner Shell, Overwrap of Carbon Fiber, Fiberglass and Epoxy Resin
    Certifications NFPA 1981 SCBA, 2002, SEI ANSI, UL 913
    • Updated SCBA meets all requirements of NFPA 1981 Open-Circuit SCBA Standard, 2002 edition
    Similar to Scott Air-Pak 2.2 (see separate description) but with your choice of high-pressure carbon-fiber cylinders providing either 30 or 60 minutes of breathable air at 4500psi. Components include: AV-3000™ facepiece; E-Z Flo™ pressure-demand regulator; Vibralert™ low-air alarm; lightweight, lumbar supported backpack; heads-up display (HUD); and a Rapid Intervention Crew/Universal Air Connection (RIC/UAC).
    System can be purchased with the Life-Link Emergency Breathing Support System (EBSS) which allows two users with similarly equipped Air-Pak products to share a common air supply. Life-Link features a tactile, quick-connect coupling on the regulator supply line. Or choose only the quick-connect regulator. Molded case can be ordered for all models.
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback