Scott™ AV-3000™ Facepieces Available on GSA/VA Contract for Federal Government customers only.
Improve on the popular AV-2000™ facepiece
Manufacturer: Scott Health & Safety 80577301
 View more versions of this product
Catalog No. 19-148-613
  Check Availability
Add to Cart
Required Accessories : Requires adapter SCT80405701.
    Specifications
    For Use With (Equipment) 42cfr84 cartridge
    Product Type Facepiece
    For Use With (Application) Used as an air purifying respirator
    Includes Larger multidirectional voicemitters
    Size Small
    Certifications NIOSH, NFPA 2007
    Design Type Simplistic clam shell
    Harness Style Kevlar
    Material (Seal) Polyisoprene
  • Description & Specifications
Related Products
Product Content Feedback