Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly Clark Professional Scott™ Essential Manual Skin Care Dispenser

Supplier:  Kimberly Clark Professional 92145

Catalog No. 17-010-540


Explore available promotions
Add to Cart

Description

Description

  • High capacity manual soap and hand sanitizer dispenser
  • Black, 1.0 L capacity
  • Dimensions 4.85 in. x 8.36 in. x 5.43 in.
  • Holds compatible skincare refills including soap, hand sanitizer, body wash, and lotion
  • Reduces run-out and maintenance time
  • Compatible with universal mounting brackets and wall and floor protector
  • Backed by a lifetime warranty and is designed to save time and money while reducing service disruptions and user complaints
Specifications

Specifications

Dispenser
Universal Mounting Brackets
Soap, Hand Sanitizer, Body Wash, Lotion
1 Ea.
Black
1 Liter
92145
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.