Scott™ Model 63 Canister-Type Gas Mask/Full-Facepiece Respirators
With Scott-O-Vista™ facepiece
Manufacturer: Scott Health & Safety 80407101
 View all options for this product
Catalog No. 19-087419
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    • Combine air-purifying protection with low breathing resistance, mobility, and minimal weight
    • Canisters are constructed of drawn aluminum bodies and strong Cycolac™ plastic tops, making them lightweight
    • Combination of new sorbent retention method and larger cross-sectional area provides greater dynamic capacity for sorbing gases and vapors
    • Features polyester harness
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback