Learn More
Scott Safety™ 4.5 Air-Pak™ 75 SCBA: E-Z Flo+ Available on GSA/VA Contract for Federal Government customers only.
Uses 4500 psig cannisters
Manufacturer:  Scott Health & Safety AP2140203200202
 View more versions of this product
Catalog No. 19-168-730
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
2 SCBA/Cs.
4.5 Air-Pak 75 SCBA
Used by first responders and municipal and industrial firefighters
Standard
E-Z Flo+
4500psi
Standard
AV-3000 Facepiece
Dual EBSS, PASS Console
NIOSH, MSHA 42CFR 84, NFPA 1981, 2007, CBRN