Scott Safety™ Air-Pak 75 SCBA Available on GSA/VA Contract for Federal Government customers only.
Offers redundant safety features, ease of use and low cost of ownership
Manufacturer: Scott Health & Safety AP2140203000102
 View more versions of this product
Catalog No. 19-500-602
  Check Availability
Add to Cart
    Specifications
    Quantity 2 SCBA/Cs.
    For Use With (Application) Used by first responders and municipal and industrial firefighters
    Facepiece Type AV-3000 Facepiece
    Hose Type Standard
    Regulator Type E-Z Flo+
    Includes Gauge and HUD Only Console
    Product Type 4.5 Air-Pak 75 SCBA
    Certifications NIOSH, MSHA 42CFR 84, NFPA 1981, 2007
    Harness Style Standard
    Pressure 4500psi
  • Description & Specifications
Related Products
Product Content Feedback