Scott Safety™ 4.5 Air-Pak™ 75 SCBA: E-Z Flo+
Uses 4500 psig cannisters
Manufacturer: Scott Health & Safety AP2140204100101
 View all options for this product
Catalog No. 19-500-650
  Check Availability
Add to Cart
    Quantity 1 SCBA/Cs.
    Pressure 4500psi
    Product Type 4.5 Air-Pak 75 SCBA
    Harness Style Standard
    For Use With (Application) Used by first responders and municipal and industrial firefighters
    Facepiece Type AV-3000 Facepiece
    Hose Type Standard
    Includes EBSS, Gauge and HUD Only Console
    Regulator Type E-Z Flow+ with Quick Connect
    Certifications NIOSH, MSHA 42CFR 84, NFPA 1981, 2007
    CBRN-approved assembly meets demands of NFPA 1981, 2007 Edition. Buddy system indicator lights and sounders indicate PASS and cylinder status to fellow firefighters; angled PASS sensor makes it easier to see console. Double-locking latch with durable para-aramid band allows quick cylinder change. Central location for batteries facilitates rapid change-out.
    Exclusive Backframe
    • Streamlined backframe promotes greater freedom of movement and reduces fatigue
    • Places bulk of SCBA weight on the hips where padded wraparound wings provide support
    • Choose parachute-style or optional alligator clip harness fasteners
    • Includes integrated donning/carrying handles and drag rescue loop
    Unique Pressure Reducer
    • Reduction in primary pressure triggers an automatic transfer to secondary circuit
    • Dual path pressure reducer ensures constant flow of air
    • Features improved hose management for less of a snag hazard
    AV-3000™ Facepiece
    • Sleek, low-profile facepiece permits better downward and peripheral vision
    • Six-point quad adjustment head harness
    • Large dual voicemitters
    • Dual mounting brackets for communication interface on either side
    EZ-Flo™ Regulator
    • Rugged, glove-friendly design
    • First breath activation makes breathing easier
    • Reduces exhalation force while sustaining airflow volumes of more than 400L/min.
    Air-Saver Switch
    • Angled for easy operation with gloved hand
    • Stops free flow of air during donning and doffing
    • Places SCBA in stand-by mode with activated cylinder
    HUD Integrated Electronics
    • Visualert™ heads-up display conveniently mounts to EZ-Flo Regulator
    • Hard-wired and engineered to ensure accurate readings
    • Unobstructed view through facepiece keeps user informed of air status in quarter-bottle increments
    EPIC™ Voice Amplifier (Optional Accessory)
    • Small-profile, simple-to-use voice projection device transmits strong, clear, crisp voice communications
    • Standalone amplifier permits hands-free communications
    Rapid Intervention Crew/Universal Air Connection (RIC/UAC)
    • Allows air to be supplied to a downed or trapped firefighter by either a Scott RIT-Pak™ II or another compressed air source
  • End of Service Indicators
    • Dual low-air alarm package provides exceptional security
    • Vibralert complements heads-up display by vibrating facepiece to ensure alarm is noticed
  • Description & Specifications
Related Products
Product Content Feedback