Scott Safety™ 4.5 Air-Pak™ 75 SCBA: E-Z Flo+ With Quick Connect Available on GSA/VA Contract for Federal Government customers only.
Uses 4500 psig cannisters
Manufacturer: Scott Health & Safety AP2240204200201
 View more versions of this product
Catalog No. 19-500-604
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Quantity 1 SCBA/Cs.
    For Use With (Application) Used by first responders and municipal and industrial firefighters
    Facepiece Type AV-3000 Facepiece
    Hose Type Quick-Connect Hose
    Regulator Type E-Z Flow+ with Quick Connect
    Includes Spring Clips Harness, Dual EBSS, PASS Console
    Product Type 4.5 Air-Pak 75 SCBA
    Certifications NIOSH, MSHA 42CFR 84, NFPA 1981, 2007
    Harness Style Spring
    Pressure 4500psi
  • Description & Specifications
Related Products
Product Content Feedback