Scott Safety™ 5.5 Air-Pak™ 75 SCBA: E-Z Flo with Quick Connect Available on GSA/VA Contract for Federal Government customers only.
5500 psi pressure
Manufacturer: Scott Health & Safety X3224022200201
 View more versions of this product
Catalog No. 19-827-300
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Quantity 1 SCBA/Cs.
    For Use With (Application) Used by first responders and municipal and industrial firefighters
    Facepiece Type AV-3000 Facepiece
    Regulator Type Quick Connect Hose
    Hose Type Quick-Connect
    Includes Spring Clip Harness, Dual EBSS, PASS Console
    Certifications NIOSH, MSHA 42CFR 84, NFPA 1981, 2007
    Harness Style Spring
    Pressure 4500psi
    Product Type 4.5 Air-Pak 75 SCBA
  • Description & Specifications
Related Products
Product Content Feedback