Scott Safety™ 5.5 Air-Pak™ 75 SCBA: E-Z Flo with Quick Connect Available on GSA/VA Contract for Federal Government customers only.
5500 psi pressure
Manufacturer: Scott Health & Safety X3215022200201
 View more versions of this product
Catalog No. 15-499-139
  Check Availability
Add to Cart
    Specifications
    Quantity 1 SCBA/Cs.
    Facepiece Type AV-3000 Facepiece
    Regulator Type Quick Connect Hose
    Includes Dual EBSS, PASS Console
    Certifications NIOSH, MSHA 42CFR 84, NFPA 1981, 2007
    Harness Style Standard
    For Use With (Application) Used by first responders and municipal and industrial firefighters
    Hose Type Quick-Connect hose
    Pressure 5500psi
    Product Type 5.5 Air-Pak 75 SCBA
  • Description & Specifications
Related Products
Product Content Feedback