Scott Safety™ 5.5 Air-Pak™ SCBA
The 5.5 cylinder is smaller and lighter—reduces fatigue, raises productivity and increases safety
Manufacturer: Scott Health & Safety AP2150204200302
 View more versions of this product
Catalog No. 19-268-865
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Quantity 2 SCBA/Cs.
    Harness Style Standard
    For Use With (Application) Used by first responders and municipal and industrial firefighters
    Facepiece Type AV-3000 Facepiece
    Hose Type Quick-Connect Hose
    Regulator Type E-Z Flo+ with Quick Connect Hose
    Includes Dual EBSS, Pak-Tracker Locator
    Product Type 5.5 Air-Pak 75 SCBA
    Certifications NIOSH, MSHA 42CFR 84, NFPA 1981, 2007
    Pressure 5500psi
  • Description & Specifications
Related Products
Product Content Feedback