Learn More
Scott Safety™ 642 Series Twin Cartridges
For use on AV-2000™ and AV-3000™, Scott-O-Vista™ and Weld-O-Vista™ full facepieces with V-bar adapter
Manufacturer:  Scott Health & Safety 80518002
 View more versions of this product
Catalog No. 19-034-474
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
30 Pack
N95 Filter
NIOSH
N95 Filter
Av-2000™ and av-3000™, scott-o-vista™ and weld-o-vista™ full facepiece with v-bar adapter
Protection against dusts, mists, solid or water-based aerosols, particulates with mmad greater than 2μ
95%