Scott Safety™ 742 Series Air-purifying Respirator Accessory, Cartridges

Easy-breathingtwin cartridges were developed to provide optimal respiratory protection in non-IDLH industrial or HAZMAT situations

Manufacturer: Scott Health & Safety 7422PD1

 View more versions of this product

Catalog No. 19-700-952

  Check Availability
Add to Cart

    Specifications

    Quantity 1 Pair/Pk.
    For Use With (Equipment) Scott 742 Series Air Purifying Respirators,Scott 742 Series Air Purifying Respirators
    Product Type Cartridge/Filter Assembly
    For Use With (Application) Non-IDLH and non-O2 deficient applications
    Cartridge Type TEDA (radioiodine), Chlorine/P100
    Filter Type Particulate aerosols, radioiodine compounds, noxious particles
    Color White/Magenta

    • Low profile, swept back cartridge design ensures optimal field of vision and allows use of communications
    • Available in a full range of NIOSH approved, chemical cartridges or combination cartridges
    • Attaches directly to XCEL Halfmask and Promask full facepiece
    • Uses Quarter turn adapter when used with AV Series Facepieces

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit