Learn More
Scott Safety™ Accessories For SEMS II Personnel Accountability System Available on GSA/VA Contract for Federal Government customers only.
For use with the SEMS II system
Manufacturer:  Scott Health & Safety 20108801
 View more versions of this product
Catalog No. 19-823-814
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Scott's SEMS II is a remarkable accountability system that puts the incident commander at every firefighter's side, displaying information vital to personal safety in volatile and dangerous situations, while providing a continuous link of communications to the command station. And with the integrated personnel locator, it provides the peace of mind that a downed or trapped firefighter can be quickly found.
RFID / Personalized Tagging. The system can be personally customized and programmable using RFID identification cards to link a firefighter to a specific Air-Pak™ SCBA. To enter their personal information, each firefighter simply swipes their pre-programmed ID card on the back of the SEMS II console. This information is transmitted to the base command computer while SEMS II is activated. Each card has three programmable fields for firefighter identification that are programmed via an RFID tag writer and proprietary software.
RFID Cards are used to link a firefighter to specific Air-Pak SCBA.
Description & Specifications
Specifications
SEMS II Mesh Gateway USB Base Station