Scott Safety™ Air Cylinder Vertical Storage Racks, ASME Units
Safe storage for breathing air cylinders
Manufacturer: Scott Health & Safety AR1404A4001
 View more versions of this product
Catalog No. 19-827-309
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Guardian or RevolveAir breating air cylinder charge stations
    Certifications/Compliance Meets ASME, DOT, TC, CRN and UN/ISO codes
    Holds 4 Cylinders (2 x 2)
    Includes Service Valve, Burst Disc and Drain Plug
    Product Type Air Cylinder Storage Rack, Cascade Plumbed
  • Description & Specifications
Related Products
Product Content Feedback