Scott Safety™ Air-Pak™ 75 SCBA
Manufacturer: Scott Health & Safety AP2120203000202
 View more versions of this product
Catalog No. 19-122-037
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Certifications NIOSH, MSHA 42CFR 84, NFPA 1981, 2007, CBRN
    Quantity 2 SCBA/cS.
    Hose Type Standard
    Harness Style Standard
    For Use With (Application) Used by first responders and municipal and industrial firefighters
    Facepiece Type AV-3000 Facepiece
    Regulator Type E-Z Flo+
    Includes PASS Console
    Pressure 2216 psi
    Product Type 2.2 Air-Pak 75 SCBA
    • Incorporates Scott Z Flo™ regulartor and Dual EBSS (Buddy Breathing) System
    • Pressure reducer acts as two reducers in one, allowing a constant supply of air, even if one gets clogged
    • Enhanced Top-Down Convertibility with AV-3000 facepiece with SureSeal
    • Lightweight, ergonomical harness places bulk of weight on hips rather than shoulders
    • Harnes incorporates quick release take-up with parachute-style hardware
    • Intergrated handle for easier carrying, donning/doffing
    • Drag Rescue Loops assits in removing downed firefighters
    • Patted and sealed electronics protect against water infiltration
    • Streamlined hose management system
    • Constant-on pressure guage light automatically dims and brightens of the HUD
    • Pak-TrackerS Fighter Locator is installed (and intitated as an option)
    Certifications
    NFPA 1981, 2007 Edition, NIOSH and CBRN
  • Description & Specifications
Related Products
Product Content Feedback