Scott Safety™ Air-Pak™ X3 SCBA Available on GSA/VA Contract for Federal Government customers only.
Designed not only to exceed the new requirements outlined in the 2013 Edition of the 1981 and 1982 standards, but also to exceed customer expectations for what an SCBA should be. End-user feedback has been incorporated in every step of the design process to deliver to the fire service the next platform of Air-Pak SCBA.
Manufacturer: Scott Health & Safety X3315021200201
 View more versions of this product
Catalog No. 15-499-135
  Check Availability
Add to Cart
    Specifications
    Certifications 1981, 2007, 2013 NFPA
    Product Type Self-Contained Breathing Apparatus
    Includes PASS
    Harness Style Standard
    For Use With (Application) Fire ground communications are enhanced with the introduction of the AV-3000™ HT facepiece, Epic 3 communication and optimally positioned "buddy" lights "external" hud
    Hose Type Standard hose
    Pressure 5500psi
    Designed with enhancements to comfort, hose and wire management, weight distribution, fire ground communications and serviceability.
    • Fire ground communications enhanced with the introduction of the AV-3000 HT facepiece, EPIC 3 Communication and optimally positioned “buddy” lights
    • “External” HUD for ease in air status updates of the team
    • Dual redundant pressure reducer
    • Vibralert tactile alarm
    Accountability features available on selected models:
    • Pak Alert PASS (Personal Alert Safety System) - standardized sound signals man-down situation
    • Pak Tracker Firefighter Locator Systems with HHR (handheld receiver) - alerts remote users of man-down situation; directional HHR provides direction for rescue teams
    • SEMS II - includes Pak Alert PASS and PAK Alert locator features plus redundant network to carry PASS and low-air notifications, reducing radio traffic
  • Description & Specifications
Related Products
Product Content Feedback