Learn More
Scott Safety™ AV-2000™ Facepiece
Interchangeable with Scott's full line of air-supplied, air-purifying, and communications products
Manufacturer:  Scott Safety™ 80401902
 View more versions of this product
Catalog No. 19-060641
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Ideal for firefighting, industrial, and hazmat service. Available in four color-coded sizes to effectively fit those with different facial configurations.
  • Cone-shaped lens provides widest possible field of vision and helps reduce carbon dioxide buildup
  • Easily disassembled for cleaning and inspection
  • Comfortable, easy-to-remove nosecup
  • Double lip design provides positive pressure to the seal
  • Broad, deep chin cup allows great contact
  • Comfort Seal™ model features sealing edge with wide, soft surface for comfort
Certifications
NIOSH approved.
Specifications
Specifications
NIOSH
Scott air supplied, air purifying, communication products
Facepiece
Large
Ideal for firefighting, industrial, hazmat service
Safety
Black
Without head harness