Scott Safety™ AV-2000™ Facepiece

Interchangeable with Scott's full line of air-supplied, air-purifying, and communications products

Manufacturer: Scott Health & Safety 80406901

 View more versions of this product

Catalog No. 19-075890

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    For Use With (Equipment) Scott air supplied, air purifying, communication products
    Product Type Respiratory Facepiece
    For Use With (Application) Ideal for firefighting, industrial, hazmat serviceIdeal for firefighting, industrial, hazmat service
    Size Small
    Certifications NIOSH
    Harness Style Polyester
    Material (Seal) EPDM Rubber
    Color Green

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit