Scott Safety™ AV-3000™ Facepiece
Builds on the success of earlier AV-Series facepieces with design enhancements for better fit, added comfort, improved field of vision and enhanced communications
Manufacturer: Scott Health & Safety 80577407
 View more versions of this product
Catalog No. 19-827-906
  Check Availability
Add to Cart
Required Accessories : To use as an air-purifying respirator, requires cartridge adapter 805622-01 for 742 Series cartridges, 805059-01 for 40mm NIOSH cartridges and 200423-01 for CBRN Cap-1 canister
    Specifications
    For Use With (Equipment) Av-3000™ facepiece
    Product Type Facepiece
    Includes Large dual voicemitters
    Size Small
    Certifications NIOSH, ANSI Z87.1-2003
    Harness Style Polyester
    Latex Free Yes
    Material (Seal) Polyisoprene
  • Description & Specifications
Related Products
Product Content Feedback