Scott Safety™ AV-3000™ Facepiece
Builds on the success of earlier AV-Series facepieces with design enhancements for better fit, added comfort, improved field of vision and enhanced communications
Manufacturer: Scott Health & Safety 80577409
 View more versions of this product
Catalog No. 19-827-908
  N/A
Add to Cart
Required Accessories : To use as an air-purifying respirator, requires cartridge adapter 805622-01 for 742 Series cartridges, 805059-01 for 40mm NIOSH cartridges and 200423-01 for CBRN Cap-1 canister
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    For Use With (Equipment) Av-3000™ facepiece
    Product Type Facepiece
    Includes Large dual voicemitters
    Size Large
    Certifications NIOSH, ANSI Z87.1-2003
    Harness Style Polyester
    Latex Free Yes
    Material (Seal) Polyisoprene
  • Description & Specifications
Related Products
Product Content Feedback