Learn More
Scott Safety™ AV-3000™ Facepiece Available on GSA/VA Contract for Federal Government customers only.
Builds on the success of earlier AV-Series facepieces with design enhancements for better fit, added comfort, improved field of vision and enhanced communications
Manufacturer:  Scott Safety™ 80577371
 View more versions of this product
Catalog No. 19-168-373
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Requires : To use as an air-purifying respirator, requires cartridge adapter 805622-01 for 742 Series cartridges, 805059-01 for 40mm NIOSH cartridges and 200423-01 for CBRN Cap-1 canister
Description
Description
  • Top-Down Convertibility allows single facepiece for APR, PAPR, SAR and SCBA respirators
  • One fit test and reduced training
  • All with Comfort Seal with deep chin cup for reliable, comfortable fit
  • Mechanical and electronic communication options across all applications
  • All components latex free
  • Four-point Kevlar™, polyester or rubber head harness options
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Fit testing required prior to using this facepiece. Prescription lens kit (805753-01) and other accessories available separately
Certifications
All head harnesses NIOSH approved for APR, supplied air, SCBA and PAPR respirators. Rubber and Kevlar head harnesses also NIOSH approved for CBRN respirators.
Compliance
All head harnesses meet ANSI Z87.1-2003. Rubber and Kevlar head harnesses also meet NFPA 1981:2007.
Specifications
Specifications
NIOSH, ANSI Z87.1-2003, NFPA 1981:2007
Polyisoprene Seal
Large dual voicemitters
Dupont™ Kevlar™ aramid fibers
Av-3000™ facepiece
Facepiece
Small
Yes