Learn More
Scott Safety™ AV-3000™ Facepiece
Builds on the success of earlier AV-Series facepieces with design enhancements for better fit, added comfort, improved field of vision and enhanced communications
Supplier:  Scott Safety™ 80577375
 View more versions of this product
Catalog No. 19-827-904
Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Requires : To use as an air-purifying respirator, requires cartridge adapter 805622-01 for 742 Series cartridges, 805059-01 for 40mm NIOSH cartridges and 200423-01 for CBRN Cap-1 canister
Description
Description
  • Top-Down Convertibility allows single facepiece for APR, PAPR, SAR and SCBA respirators
  • One fit test and reduced training
  • All with Comfort Seal with deep chin cup for reliable, comfortable fit
  • Mechanical and electronic communication options across all applications
  • All components latex free
  • Four-point Kevlar™, polyester or rubber head harness options
Order Info
Fit testing required prior to using this facepiece. Prescription lens kit (805753-01) and other accessories available separately
Certifications
All head harnesses NIOSH approved for APR, supplied air, SCBA and PAPR respirators. Rubber and Kevlar head harnesses also NIOSH approved for CBRN respirators.
Compliance
All head harnesses meet ANSI Z87.1-2003. Rubber and Kevlar head harnesses also meet NFPA 1981:2007.
Specifications
Specifications
NIOSH, ANSI Z87.1-2003, NFPA 1981:2007
Facepiece
Av-3000™ facepiece
Yes
Dupont™ Kevlar™ aramid fiber, Polyisoprene Seal
Medium
Communication bracket and large dual voicemitters