Scott Safety™ AV-Hood™

Unique fastening system facilitates quick donning and ensures proper hood/face interface

Manufacturer: Scott Health & Safety 805289-05

 View more versions of this product

Catalog No. 19-065-301

  Check Availability
Add to Cart

    Specifications

    Color Gold
    Product Type Hood
    For Use With (Application) Unique fastening system facilitates quick donning and ensures proper hood/face interface
    Includes Installation kit
    Material PBI
    Certifications NFPA 1971

    • Easily assembled and disassembled for routine cleaning and maintenance
    • Choose from three flame-resistant fabric options

    Compliance

    Meets all NFPA 1971 requirements

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit