Scott Safety™ Eagle Imager™ TCS™ Truck-Mount Charging System

Structurally designed system allows both trickle charging and smart charging

Manufacturer: Scott Health & Safety 805621-02

 View more versions of this product

Catalog No. 19-027-342

  Check Availability
Add to Cart

    Specifications

    Voltage 110V
    For Use With (Equipment) Eagle imager I thermal imaging camera
    Product Type Truck mount charging system

    • Choose from models with 110VAC or 12VDC cigarette-style adapter
    • Movable storage clip for pistol grip
    • Spring loaded locking mechanism ensures proper restraint of camera
    • Charges fully depleted spare battery in two hours
    • Can be mounted in horizontal or vertical position
    • Withstands 9-G of shock, as required by NFPA for objects mounted in fire service vehicles

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit