Scott Safety™ Envoy Radiocom™
Ideal communications system for first responders builds on the features of the E-Z Radiocom
Manufacturer: Scott Health & Safety 80526203
 View more versions of this product
Catalog No. 19-818-685
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Communications system suits volunteer firefighters, EMS personnel, Haz Mat responders, and industrial users. Many more models are available for different radio systems; please contact your Fisher Safety Representative for more information.
    ·    Features a facepiece-mounted microphone, ear speaker boom and a chest-mounted Push-to-Talk (PTT) console
    ·    Chest-mounted PTT console may be used independently as a lapel microphone in nontactical applications
  • Description & Specifications
Related Products
Product Content Feedback